missing translation for 'onlineSavingsMsg'
Learn More

Mannose triflate, 98%

Product Code. 15587049 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
100 mg
1 g
Unit Size:
100mg
1g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15587049 100 mg 100mg
15577049 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15587049 Supplier Thermo Scientific Acros Supplier No. 461341000
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 92051-23-5 | C15H19F3O12S | 480.36 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 92051-23-5
Molecular Formula C15H19F3O12S
Molecular Weight (g/mol) 480.36
InChI Key OIBDVHSTOUGZTJ-PEBLQZBPSA-N
Synonym tatm, mannose triflate, 1,3,4,6-tetra-o-acetyl-2-o-trifluoromethanesulfonyl-beta-d-mannopyranose, 1,3,4,6-tetra-o-acetyl-2-o-trifluoromethanesulfonyl-b-d-mannopyranose, unii-0ki8ffq6tt, 0ki8ffq6tt, beta-d-mannopyranose 1,3,4,6-tetra-o-acetate 2-o-trifluoromethanesulfonate, 2r,3r,4s,5s,6s-3,4,6-tris acetyloxy-5-trifluoromethanesulfonyloxy oxan-2-yl methyl acetate, mannose trilfate
PubChem CID 11113597
IUPAC Name [(2R,3R,4S,5S,6S)-3,4,6-triacetyloxy-5-(trifluoromethylsulfonyloxy)oxan-2-yl]methyl acetate
SMILES CC(=O)OCC1C(C(C(C(O1)OC(=O)C)OS(=O)(=O)C(F)(F)F)OC(=O)C)OC(=O)C

Specifications

Melting Point 118.0°C to 122.0°C
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (ELSD) (HPLC)
Quantity 100 mg
Solubility Information Clear colorless to slightly yellow
Formula Weight 480.36
Percent Purity 98%
Chemical Name or Material Mannose triflate

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.