missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Mannose Phosphate Isomerase Antibody (OTI4G7), FITC, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-72584F
This item is not returnable.
View return policy
Description
Mannose Phosphate Isomerase Monoclonal specifically detects Mannose Phosphate Isomerase in Human, Rat samples. It is validated for Flow Cytometry, Immunohistochemistry.
Specifications
| Mannose Phosphate Isomerase | |
| Monoclonal | |
| FITC | |
| PBS with 0.05% Sodium Azide | |
| MPI | |
| Full length recombinant protein of human Mannose Phosphate Isomerase (NP_002426) produced in HEK293T cells. | |
| 0.1 mL | |
| Primary | |
| Human, Rat | |
| IgG1 |
| Flow Cytometry, Immunohistochemistry | |
| OTI4G7 | |
| Flow Cytometry, Immunohistochemistry | |
| CDG1B, EC 5.3.1, EC 5.3.1.8, FLJ39201, mannose phosphate isomerase, mannose-6- phosphate isomerase, mannose-6-phosphate isomerase, Phosphohexomutase, Phosphomannose isomerase, phosphomannose isomerase 1, PMI, PMI1phosphohexomutase | |
| Mouse | |
| Affinity Purified | |
| Lipid and Metabolism | |
| 4351.0 | |
| Store at 4°C in the dark. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction