missing translation for 'onlineSavingsMsg'
Learn More

Mannitol, Mikromol™

Product Code. 17969104
Change view
Click to view available options
Quantity:
250 mg
Unit Size:
250mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
17969104 250 mg 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 17969104 Supplier Mikromol Supplier No. MM0654.00
Big Savings for Better Separation
Use promo code 27992
View all promotions
 Restricted Item
Estimated Shipment: 30-10-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.

Mannitol
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material D-Mannitol
CAS 69-65-8
Analyte or Component Names D-Mannitol
Shipping Condition Room Temperature
Molecular Formula C6 H14 O6
Certifications/Compliance ISO 17034
Synonym Mannitol, d-Mannitol, D-Mannitol, Cordycepic acid (6CI,7CI), Mannitol, D- (8CI), Brightmoon, Bronchitol, Cerestar 16700, D-(-)-Mannitol, D-Mannit, Diosmol, E 421, Isotol, Manicol, Maniton S, Manna sugar, Mannidex, Mannidex 16700, Mannigen, Mannistol, Mannit, Mannit 60, Mannit P, Mannit S, Mannite, Mannitol 35, Mannitol 60, Mannitol P 60, Mannitol SD 200, Mannitolum, Mannogem 2080, Mannogem EZ, Marine Crystal, Nonpareil 108, Nonpareil 108(200), Orocell 200, Osmitrol, Osmosal, Parteck 300, Parteck Delta M, Parteck M 100, Parteck M 200, Parteck M 300, Partek M, Pearlitol, Pearlitol 100, Pearlitol 100SD, Pearlitol 160C, Pearlitol 2005D, Pearlitol 200SD, Pearlitol 25C, Pearlitol 50C, Pearlitol SD 100, Perlitol SD 200, Perteck M, PharmMannidex, PharmMannidex 16701, Resectisol, Lactitol Monohydrate Impurity C, Sorbitol Impurity A, myo-Inositol Impurity A, Lactitol Impurity C
InChI Formula InChI=1S/C6H14O6/c7-1-3(9)5(11)6(12)4(10)2-8/h3-12H,1-2H2/t3-,4-,5-,6-/m1/s1
SMILES OC[C@@H](O)[C@@H](O)[C@H](O)[C@H](O)CO
IUPAC Name (2R,3R,4R,5R)-hexane-1,2,3,4,5,6-hexol
Molecular Weight (g/mol) 182.17
Quantity 250 mg
Formula Weight 182.079 g/mol
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.