missing translation for 'onlineSavingsMsg'
Learn More

Mannitol, ACS Reagent Grade, Ricca Chemical

Product Code. 31750721
Change view
Click to view available options
Quantity:
500 g
2.5 kg
Unit Size:
2.50 L
500g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
31750721 500 g 500g
31750697 2.5 kg 2.50 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 31750721 Supplier Ricca Chemical Company Supplier No. RDCM0200500B1
 Restricted Item
Estimated Shipment: 05-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Chemical Identifiers

CAS 69-65-8
Molecular Formula C6H14O6
Molecular Weight (g/mol) 182.17
MDL Number MFCD00064287
InChI Key FBPFZTCFMRRESA-UHFFFAOYNA-N
IUPAC Name hexane-1,2,3,4,5,6-hexol
SMILES OCC(O)C(O)C(O)C(O)CO

Specifications

Boiling Point Approximately 290°C to 295°C
Ignition Residue max. 0.01%
Loss on Drying max. 0.05%
Packaging White Poly Wide Mouth Bottle
Quantity 500 g
Specific Rotation 23.3-24.3°
Grade ACS
Insoluble Matter max. 0.01%
Titratable Acid max. 0.0008 meq/g
Physical Form Crystalline Powder or Lumps
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.