missing translation for 'onlineSavingsMsg'
Learn More

Mangiferin, TRC

Product Code. 30133964
Change view
Click to view available options
Quantity:
100 mg
250 mg
1 g
Unit Size:
100mg
1g
250mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30133964 250 mg 250mg
30137060 1 g 1g
30005412 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30133964 Supplier Toronto Research Chemicals Supplier No. TRCM163800250MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 4773-96-0
Molecular Formula C19 H18 O11
Molecular Weight (g/mol) 422.34
Synonym 9H-Xanthen-9-one, 2-β-D-glucopyranosyl-1,3,6,7-tetrahydroxy-, Mangiferin (6CI,7CI,8CI), 1,3,6,7-Tetrahydroxyxanthone-C2-β-D-glucoside, 2-C-β-D-Glucopyranosyl-1,3,6,7-tetrahydroxyxanthone, 2-β-D-Glucopyranosyl-1,3,6,7-tetrahydroxy-9H-xanthen-9-one, Alpizarin, Alpizarine, Aphloiol, Chinomin, Chinonin, Hedysarid, NSC 248870, Shamimin
IUPAC Name 1,3,6,7-tetrahydroxy-2-[(2S,3R,4R,5S,6R)-3,4,5-trihydroxy-6-(hydroxymethyl)oxan-2-yl]xanthen-9-one
SMILES OC[C@H]1O[C@H]([C@H](O)[C@@H](O)[C@@H]1O)c2c(O)cc3Oc4cc(O)c(O)cc4C(=O)c3c2O

Specifications

Quantity 250 mg
InChI Formula InChI=1S/C19H18O11/c20-4-11-15(25)17(27)18(28)19(30-11)12-8(23)3-10-13(16(12)26)14(24)5-1-6(21)7(22)2-9(5)29-10/h1-3,11,15,17-23,25-28H,4H2/t11-,15-,17+,18-,19+/m1/s1
Formula Weight 422.0849
Percent Purity >95%
Chemical Name or Material Mangiferin
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.