missing translation for 'onlineSavingsMsg'
Learn More

Manganous Sulfate, MnSO4-H2O, MP Biomedicals

Product Code. 11408845 Shop All MP Biomedicals Products
Change view
Click to view available options
Quantity:
100 g
Unit Size:
100g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11408845 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11408845 Supplier MP Biomedicals Supplier No. 0522509980
 Restricted Item
Estimated Shipment: 22-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity manganese(II) sulfate monohydrate used in diverse biological research and industrial applications.

Specifications

CAS 10034-96-5
Physical Form Powder
Quantity 100 g
Molecular Formula H2MnO5S
MDL Number MFCD00149159
Synonym manganese sulfate monohydrate, manganese ii sulfate monohydrate, manganese sulfate hydrate, manganese ii sulfate hydrate, mnso4.h2o, manganous sulfate monohydrate, unii-w00lys4t26, manganese 2+ sulfate monohydrate, manganese sulfate usp, manganese 2+ sulfate hydrate
InChI Key ISPYRSDWRDQNSW-UHFFFAOYSA-L
SMILES O.[Mn++].[O-]S([O-])(=O)=O
IUPAC Name manganese(2+) hydrate sulfate
Molecular Weight (g/mol) 169.01
PubChem CID 177577
ChEBI CHEBI:86364
Formula Weight 223.06
Grade Research
Chemical Name or Material Manganous Sulfate
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • Manganous sulfate, hydrate, 100 g
Signal Word
  • Warning
Hazard Category
  • LONG-TERM AQUATIC HAZARD Chronic 2
  • Specific target organ toxicity after repeated exposure Category 2
Hazard Statement
  • H373-May cause damage to organs.
  • H411-Toxic to aquatic life with long lasting effects.
Precautionary Statement
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P273-Avoid release to the environment.
  • P314-Get medical advice/attention if you feel unwell.
  • P391-Collect spillage.
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Supplemental information
  • MIXTURE LIST-Contain: Manganese sulphate hydrate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.