missing translation for 'onlineSavingsMsg'
Learn More
Learn More
WTW™ Manganese Cell Test Kits
Click to view available options
No. of Tests:
25
Unit Size:
Pack of 25
Description
- Convenient and cost-effective
- Assured quality by AQA/IQC
- For infrequent or very large sample throughput in the laboratory
- Sample adding, inserting, measuring and reading
- Reaction cuvette with barcode or method selection
Specifications
Specifications
| Analytes | Formaldoxime |
| For Use With (Application) | For waste water, drinking water |
| For Use With (Equipment) | PhotoLab™ S6, PhotoLab™ S12, PhotoLab™ Spectral, Photoflex photometers |
| Includes | Certificates |
| No. of Tests | 25 |
| Range | 0.10 - 5.00mg/L |
| Sufficient For | 25 Tests |
| Test Method | Photometric |
| Type | CombiCheck Manganese (Mn) cell test kit |
Safety and Handling
Product Identifier
- Manganese Cell Test Method: photometric 0.10 - 5.00 mg/l Mn Spectroquant
- Danger
- Acute toxicity Category 4
- Acute toxicity Category 3
- Skin corrosion/irritation Category 1 B
- Specific target organ toxicity Category 3
- LONG-TERM AQUATIC HAZARD Chronic 3
- Substances and mixtures corrosive to metals Category 1
- Skin corrosion/irritation Category 2
- Serious eye damage/eye irritation Category 2
- Skin sensitiser Category 1
- H290-May be corrosive to metals.
- H302-Harmful if swallowed.
- H311-Toxic in contact with skin.
- H314-Causes severe skin burns and eye damage.
- H315-Causes skin irritation.
- H317-May cause an allergic skin reaction.
- H319-Causes serious eye irritation.
- H335-May cause respiratory irritation.
- H351-Suspected of causing cancer.
- H373-May cause damage to organs.
- H412-Harmful to aquatic life with long lasting effects.
- P273-Avoid release to the environment.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
- P302+P352-IF ON SKIN: Wash with plenty of water/soap.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P308+P311-IF exposed or concerned: Call a POISON CENTER/doctor/.
- MIXTURE LIST-Contains : ammonia solution, Potassium cyanide; Hydroxylammonium chloride; formaldehyde, methanol
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction