missing translation for 'onlineSavingsMsg'
Learn More

Manganese telluride, 99.9% (metals basis)

Product Code. 11347848
Change view
Click to view available options
Quantity:
10 g
50 g
Unit Size:
10g
50g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11347848 50 g 50g
11357848 10 g 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11347848 Supplier Thermo Scientific Alfa Aesar Supplier No. 036389.18
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

CAS: 12032-88-1 | MnTe | 182.54 g/mol

Manganese telluride is used in scientific research and developments.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 12032-88-1
Molecular Formula MnTe
Molecular Weight (g/mol) 182.54
MDL Number MFCD00049487
InChI Key VMINMXIEZOMBRH-UHFFFAOYSA-N
Synonym manganese telluride, telluroxomanganese, manganese monotelluride, manganous telluride, manganese ii telluride, manganese 2+ telluride
PubChem CID 82828
IUPAC Name tellanylidenemanganese
SMILES [Mn]=[Te]

Specifications

Color Gray
Physical Form Lump
Quantity 50 g
Formula Weight 182.54
Percent Purity 99.9%
Assay (metals basis)
Chemical Name or Material Manganese telluride

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.