missing translation for 'onlineSavingsMsg'
Learn More

Manganese Sulfate Monohydrate, TRC

Product Code. 30138149
Change view
Click to view available options
Quantity:
1 g
10 g
50 g
Unit Size:
10g
1g
50g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30138149 10 g 10g
30137488 50 g 50g
30008300 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30138149 Supplier Toronto Research Chemicals Supplier No. TRCM16432010G
 Restricted Item
Estimated Shipment: 11-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 10034-96-5
Molecular Formula Mn . O4 S . H2 O
Molecular Weight (g/mol) 169.02
Synonym Manganese sulfate monohydrate, Manganese Mesosulfate (MnSO4.H2O), Sulfuric Acid Manganese(2+) Salt Monohydrate, Manganese Monosulfate Monohydrate, Manganese Sulfate (MnSO4) Monohydrate, Manganese Sulfate Monohydrate, Manganese(2+) Sulfate Monohydrate, Manganese(II) Sulfate Monohydrate, Manganous Sulfate Monohydrate, Sulfuric Acid Manganese salt (1:1) Monohydrate
IUPAC Name manganese(2+);sulfate;hydrate
SMILES O.[Mn+2].[O-]S(=O)(=O)[O-]

Specifications

Quantity 10 g
InChI Formula InChI=1S/Mn.H2O4S.H2O/c;1-5(2,3)4;/h;(H2,1,2,3,4);1H2/q+2;;/p-2
Formula Weight 168.9003
Chemical Name or Material Manganese Sulfate (AS)
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.