missing translation for 'onlineSavingsMsg'
Learn More

Manganese Standard for Ion Chromatography (IC) 1 mg/mL (1000 ppm) in 0.005% Nitric Acid (HNO3), Reagecon
BIOPHARMA

Product Code. 11556683 Shop All Reagecon Products
Change view
Click to view available options
Quantity:
100 mL
Unit Size:
100mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11556683 100 mL 100mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11556683 Supplier Reagecon Supplier No. ICCS11

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Manufactured gravimetrically from ultra-pure salt, tested by an ISO17025 Accredited Test Method (INAB Ref:264T), and assayed by titration and ICP-MS prior to manufacture.

  • Ion Chromatography Reference Standard, Ion Chromatography Reference Material
  • ISO/IEC 17025 for the testing of ICP/ICP-MS Standards using ICP-MS (INAB Ref: 264T)
  • ISO/IEC 17025 for calibration of laboratory balances (INAB Ref: 265C)
  • ISO/IEC 17025 for temperature calibration in the range of -196 - +1200°C (INAB Ref: 265C)
  • ISO/IEC 17025 for density measurement (INAB Ref: 264T) using Oscillating U-Tube and Bingham Pycnometry
  • ISO/IEC 17025 for certain complexometric titrations (INAB Ref: 264T)
  • GHS/OSHA compliant, REACH etc.
  • This product is produced in a highly controlled cleanroom (ISO 7) environment, using ultra-pure water (specially treated for the production of ion chromatography standards) and triple leached pre-cleaned bottles.
  • The titrimetry and spectroscopy are controlled and calibrated using separate Certified Reference Materials (CRM’s), so the product comes with two layers of traceability including NIST Standards where available.
  • All of the uncertainties of measurement for the product are calculated according to Eurachem/CITAC guidelines and reported as expanded uncertainties at the 95% confidence level.

Specifications

Description Matrix: 0.005% HNO₃
Packaging HDPE Triple-leached
Analyte or Component Names Manganese
Physical Form Liquid
Concentration 1 mg/mL (1000 ppm)
Technique Ion Chromatography, Cation Measurement
Element Mn
Quantity 100 mL
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.