missing translation for 'onlineSavingsMsg'
Learn More

Manganese Standard for ICP/MS - Claritas PPT Grade, SPEX CertiPrep™

Product Code. 15255946 Shop All SPEX Certiprep Products
Click to view available options
Quantity:
125 mL
Unit Size:
125mL
This item is not returnable. View return policy

Product Code. 15255946

Brand: SPEX Certiprep CLMN21BY

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Specifications

CAS 7439-96-5
Molecular Formula Mn
MDL Number MFCD00011111
InChI Key PWHULOQIROXLJO-UHFFFAOYSA-N
SMILES [Mn]
IUPAC Name manganese
Solvent or Matrix 2% HNO3
Molecular Weight (g/mol) 54.94
PubChem CID 23930
Quantity 125 mL
ChEBI CHEBI:35154
Concentration or Composition (by Analyte or Components) 1mg/L
Chemical Name or Material Manganese Standard for ICP/MS - Claritas PPT Grade
Show More Show Less
compliance-icons
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
Precautionary Statement
  • P264-Wash thoroughly after handling.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P321-Specific treatment (see on this label).
  • P332+P313-If skin irritation occurs: Get medical advice/attention.
  • P337+P313-If eye irritation persists: Get medical advice/attention.
  • P362+P364-Take off contaminated clothing and wash it before reuse.
Supplemental information
  • MIXTURE LIST-Contains : nitric acid
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.