missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Manganese Standard, 1000mg/L in 2% HNO3 (Assurance Grade), SPEX CertiPrep™
Shop All SPEX Certiprep ProductsDescription
Inorganic Standard manufactured for AA, ICP, ICP/MS, IC, XRF, and other analytical instrumentation
- Manganese Standard
- Made with ASTM Type I Water
- 1,000 μg/mL concentration
- 250 mL volume
Certifications
ISO/IEC 17025:2005, ISO 17034:2016, ISO 9001: 2015
Specifications
Specifications
| Packaging | Poly Bottle |
| Certificate of Analysis | Assurance Certificate of Analysis reports up to 30 elements at ppb levels. |
| CAS Min % | 0.1, 2.0, 97.9 |
| CAS Max % | 0.1, 2.0, 97.9 |
| Technique | AA, ICP |
| Type | Certified Reference Material |
| Quantity | 250 mL |
| Grade | Assurance |
| Chemical Name or Material | Manganese Standard |
Safety and Handling
Product Identifier
- 1000 µg/ml manganese
- Warning
- Serious eye damage/eye irritation Category 2
- Skin corrosion/irritation Category 2
- H315-Causes skin irritation.
- H319-Causes serious eye irritation.
- P264-Wash thoroughly after handling.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P332+P313-If skin irritation occurs: Get medical advice/attention.
- P361+P364-Take off immediately all contaminated clothing and wash it before reuse.
- P337+P313-If eye irritation persists: Get medical advice/attention.
- MIXTURE LIST-Contain: nitric acid
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?