missing translation for 'onlineSavingsMsg'
Learn More

Manganese powder, -325 mesh, 99.3% (metals basis)

Product Code. 11381298
Change view
Click to view available options
Quantity:
25 g
250 g
1 kg
Unit Size:
1kg
250g
25g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11381298 250 g 250g
11301308 25 g 25g
11391298 1 kg 1kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11381298 Supplier Thermo Scientific Alfa Aesar Supplier No. 010238.30
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Form: Apparent density 2.6-3.5g/cm3 | CAS: 7439-96-5 | Mn | 54.94 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7439-96-5
Molecular Formula Mn
Molecular Weight (g/mol) 54.94
MDL Number MFCD00011111
InChI Key PWHULOQIROXLJO-UHFFFAOYSA-N
Synonym colloidal, mangan, manganese, elemental, cutaval, magnacat, metal alloy, tronamang, fume, mangan polish, compounds
PubChem CID 23930
ChEBI CHEBI:35154
IUPAC Name manganese
SMILES [Mn]

Specifications

Color Gray
Appearance Gray powder
Quantity 250 g
Mesh Size -325 Mesh
Assay Percent Range 99.3% (Metals basis)
Form Powder
Chemical Name or Material Manganese
compliance-icons
Product Identifier
  • Manganese powder
Signal Word
  • Warning
Hazard Category
  • Flammable solid Category 2
Hazard Statement
  • H228-Flammable solid.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P370+P378-In case of fire: Use dry sand, dry chemical or alcohol-resistant foam to extinguish.
Supplemental information
  • MIXTURE LIST-Contain: Manganese

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.