missing translation for 'onlineSavingsMsg'
Learn More

Manganese(IV) telluride, 99.9% (metals basis), Thermo Scientific Chemicals

Catalog Number 11392228
Click to view available options
Quantity:
10 g
50 g
This item is not returnable. View return policy
12032-89-2
MnTe2
310.138
MFCD00064703
KDJKBQYOEWVJGP-UHFFFAOYSA-N
manganese ditelluride, manganese iv telluride, bis tellanylidene manganese, ditellanylidenemanganese, manganese telluride
82829
bis(tellanylidene)manganese
[Mn](=[Te])=[Te]
This item is not returnable. View return policy
12032-89-2
MnTe2
310.138
MFCD00064703
KDJKBQYOEWVJGP-UHFFFAOYSA-N
manganese ditelluride, manganese iv telluride, bis tellanylidene manganese, ditellanylidenemanganese, manganese telluride
82829
bis(tellanylidene)manganese
[Mn](=[Te])=[Te]

It is used as an organic intermediates.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Physical Form Pieces
Quantity 50 g
Solubility Information Insoluble in water.
Formula Weight 310.14
Percent Purity 99.9%
Odor Odorless
Assay (metals basis)
Chemical Name or Material Manganese(IV) telluride

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'