missing translation for 'onlineSavingsMsg'
Learn More

Manganese(IV) telluride, 99.9% (metals basis)

Product Code. 11302238
Click to view available options
Quantity:
10 g
50 g
Unit Size:
10g
50g
This item is not returnable. View return policy

Product Code. 11302238

Brand: Thermo Scientific Alfa Aesar 013134.09

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 12032-89-2 | MnTe2 | 310.138 g/mol

It is used as an organic intermediates.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 12032-89-2
Molecular Formula MnTe2
Molecular Weight (g/mol) 310.138
MDL Number MFCD00064703
InChI Key KDJKBQYOEWVJGP-UHFFFAOYSA-N
Synonym manganese ditelluride, manganese iv telluride, bis tellanylidene manganese, ditellanylidenemanganese, manganese telluride
PubChem CID 82829
IUPAC Name bis(tellanylidene)manganese
SMILES [Mn](=[Te])=[Te]

Specifications

Physical Form Pieces
Quantity 10 g
Solubility Information Insoluble in water.
Formula Weight 310.14
Percent Purity 99.9%
Odor Odorless
Assay (metals basis)
Chemical Name or Material Manganese(IV) telluride

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.