missing translation for 'onlineSavingsMsg'
Learn More

Manganese(Iv) Oxide, Honeywell

Product Code. p-7071932 Shop All Honeywell Research Chemicals Products
Click to view available options
Quantity:
100 g
500 g
Unit Size:
100g
500g
This item is not returnable. View return policy

Product Code. 15622620

Brand: Honeywell Research Chemicals 243442100G

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Chemical Identifiers

CAS 1313-13-9
Molecular Formula MnO2
Molecular Weight (g/mol) 86.94
MDL Number MFCD00003463
InChI Key NUJOXMJBOLGQSY-UHFFFAOYSA-N
Synonym manganese dioxide, manganese iv oxide, manganese peroxide, manganese superoxide, manganese black, manganese binoxide, manganese oxide, mangandioxid, black manganese oxide, braunstein
PubChem CID 14801
IUPAC Name dioxomanganese
SMILES O=[Mn]=O

Specifications

Quantity 100 g
UN Number NONH for all modes of transport
Formula Weight 86.94g/mol
Percent Purity ≥99%
Grade ReagentPlus™
Chemical Name or Material Manganese(Iv) Oxide
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.