missing translation for 'onlineSavingsMsg'
Learn More

Manganese(IV) oxide, 99.9% (metals basis), Thermo Scientific Chemicals

Catalog Number 11339907
Click to view available options
Quantity:
250 g
1 kg
1313-13-9
MnO2
86.94
MFCD00003463
NUJOXMJBOLGQSY-UHFFFAOYSA-N
manganese dioxide, manganese iv oxide, manganese peroxide, manganese superoxide, manganese black, manganese binoxide, manganese oxide, mangandioxid, black manganese oxide, braunstein
14801
dioxomanganese
O=[Mn]=O
1313-13-9
MnO2
86.94
MFCD00003463
NUJOXMJBOLGQSY-UHFFFAOYSA-N
manganese dioxide, manganese iv oxide, manganese peroxide, manganese superoxide, manganese black, manganese binoxide, manganese oxide, mangandioxid, black manganese oxide, braunstein
14801
dioxomanganese
O=[Mn]=O

It is used as an oxidizing agent in organic synthesis such as oxidation of allylic/benzylic alcohols, as a textile dye, as a reducing agent, and as a component of dry cell batteries such as zinc-carbon battery and alkaline battery. It is also used in making pigments for glasses and ceramics, and drier for paints. Further it is used in the manufacture of manganese steel and several manganese derivatives including potassium permanganate, a powerful versatile oxidant. Catalytic hydration of nitriles to amides by flowing through manganese dioxide has been reported. Manganese dioxide supported on inorganic oxide can be used for oxidation of methylamine through CWAO (Catalytic Wet Air Oxidation). It has high potential as highly efficient and robust material for water oxidation reaction (WORs).

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Melting Point 535°C (decomposition)
Physical Form Powder
Quantity 250 g
Assay Percent Range 99.9% (metals basis)
Merck Index 14,5730
Solubility Information Insoluble in water.
Formula Weight 86.94
Percent Purity 99.9%
Density 5.026 g/mL
Assay (metals basis)
Chemical Name or Material Manganese(IV) oxide
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • Manganese(IV) oxide
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 4
  • Specific target organ toxicity after repeated exposure Category 2
Hazard Statement
  • H302-Harmful if swallowed.
  • H332-Harmful if inhaled.
  • H373-May cause damage to organs.
Precautionary Statement
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P264-Wash face, hands, and anyexposed skin thoroughly after handling.
  • P270-Do not eat, drink or smoke when using this product.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.
Supplemental information
  • MIXTURE LIST-Contains : manganese dioxide

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'