missing translation for 'onlineSavingsMsg'
Learn More

Manganese(IV) oxide, 99.9% (metals basis)

Product Code. 11349907
Change view
Click to view available options
Quantity:
250 g
1 kg
Unit Size:
1kg
250g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11349907 1 kg 1kg
11339907 250 g 250g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11349907 Supplier Thermo Scientific Alfa Aesar Supplier No. 042250.A1
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 1313-13-9 | MnO2 | 86.94 g/mol

It is used as an oxidizing agent in organic synthesis such as oxidation of allylic/benzylic alcohols, as a textile dye, as a reducing agent, and as a component of dry cell batteries such as zinc-carbon battery and alkaline battery. It is also used in making pigments for glasses and ceramics, and drier for paints. Further it is used in the manufacture of manganese steel and several manganese derivatives including potassium permanganate, a powerful versatile oxidant. Catalytic hydration of nitriles to amides by flowing through manganese dioxide has been reported. Manganese dioxide supported on inorganic oxide can be used for oxidation of methylamine through CWAO (Catalytic Wet Air Oxidation). It has high potential as highly efficient and robust material for water oxidation reaction (WORs).

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1313-13-9
Molecular Formula MnO2
Molecular Weight (g/mol) 86.94
MDL Number MFCD00003463
InChI Key NUJOXMJBOLGQSY-UHFFFAOYSA-N
Synonym manganese dioxide, manganese iv oxide, manganese peroxide, manganese superoxide, manganese black, manganese binoxide, manganese oxide, mangandioxid, black manganese oxide, braunstein
PubChem CID 14801
IUPAC Name dioxomanganese
SMILES O=[Mn]=O

Specifications

Melting Point 535°C (decomposition)
Color Black
Physical Form Powder
Quantity 1 kg
Assay Percent Range 99.9% (metals basis)
Merck Index 14,5730
Solubility Information Insoluble in water.
Formula Weight 86.94
Percent Purity 99.9%
Density 5.026 g/mL
Assay (metals basis)
Chemical Name or Material Manganese(IV) oxide
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • Manganese(IV) oxide
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Specific target organ toxicity after repeated exposure Category 2
Hazard Statement
  • H302-Harmful if swallowed.
  • H332-Harmful if inhaled.
  • H373-May cause damage to organs.
Precautionary Statement
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P264-Wash face, hands and anyexposed skin thoroughly after handling.
  • P270-Do not eat, drink or smoke when using this product.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.
Supplemental information
  • MIXTURE LIST-Contain: Manganese dioxide

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.