missing translation for 'onlineSavingsMsg'
Learn More

Manganese(IV) oxide, 97%

Product Code. 11479662
Change view
Click to view available options
Quantity:
10 g
500 g
2500 g
10 kg
Unit Size:
10g
10kg
2500g
500g
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11479662 2500 g 2500g
11488830 10 g 10g
11469662 500 g 500g
15463027 10 kg 10kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 11479662 Supplier Thermo Scientific Alfa Aesar Supplier No. A10765.0E
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 1313-13-9 | MnO2 | 86.94 g/mol

Manganese(IV) oxide is used as a pigment and as a reagent inorganic synthesis especially for the oxidation of allylic alcohols. It is also used for dry -cell batteries such as the alkaline battery and the zinc-carbon battery. It is involved in the preparation of other manganese compounds. It is a precursor to ferromanganese and related alloys, which is useful in the steel industry. It catalyses the decomposition of hydrogen peroxide into oxygen and water. Further, it is used to prepare manganese(II) sulfate, manganese(II) chloride and potassium permanganate. In addition to this, it is used in ceramics and glass manufacturing as an inorganic pigment.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Manganese(IV) oxide is used as a pigment and as a reagent inorganic synthesis especially for the oxidation of allylic alcohols. It is also used for dry -cell batteries such as the alkaline battery and the zinc-carbon battery. It is involved in the preparation of other manganese compounds. It is a precursor to ferromanganese and related alloys, which is useful in the steel industry. It catalyses the decomposition of hydrogen peroxide into oxygen and water. Further, it is used to prepare manganese(II) sulfate, manganese(II) chloride and potassium permanganate. In addition to this, it is used in ceramics and glass manufacturing as an inorganic pigment.

Solubility
Insoluble in water.

Notes
Incompatible with strong acids, strong reducing agents and organic materials.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1313-13-9
Molecular Formula MnO2
Molecular Weight (g/mol) 86.94
MDL Number MFCD00003463
InChI Key NUJOXMJBOLGQSY-UHFFFAOYSA-N
Synonym manganese dioxide, manganese iv oxide, manganese peroxide, manganese superoxide, manganese black, manganese binoxide, manganese oxide, mangandioxid, black manganese oxide, braunstein
PubChem CID 14801
IUPAC Name dioxomanganese
SMILES O=[Mn]=O

Specifications

Melting Point 535°C (decomposition)
Quantity 2500 g
Merck Index 14,5730
Solubility Information Insoluble in water.
Formula Weight 86.94
Percent Purity 98%
Density 5.026
Chemical Name or Material Manganese (IV) oxide

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.