missing translation for 'onlineSavingsMsg'
Learn More

Manganese(III) phosphate hydrate, 99%

Product Code. 11413478
Change view
Click to view available options
Quantity:
10 g
50 g
Unit Size:
10g
50g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11413478 50 g 50g
11403478 10 g 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11413478 Supplier Thermo Scientific Alfa Aesar Supplier No. A17868.18
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 104663-56-1 | MnO4P | 149.91 g/mol

It is employed as intermediate for pharmaceutical.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is employed as intermediate for pharmaceutical.

Solubility
Insoluble in water.

Notes
Store in a tightly closed container in a cool, dry, well-ventilated area. Store away from strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 104663-56-1
Molecular Formula MnO4P
Molecular Weight (g/mol) 149.91
MDL Number MFCD00209658
InChI Key BECVLEVEVXAFSH-UHFFFAOYSA-K
PubChem CID 71317435
SMILES [Mn+3].[O-]P([O-])([O-])=O

Specifications

Quantity 50 g
Solubility Information Insoluble in water.
IUPAC Name manganese(3+) phosphate
Formula Weight 149.91 (Anhydrous)
Percent Purity 99%
Chemical Name or Material Manganese(III) phosphate hydrate

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.