missing translation for 'onlineSavingsMsg'
Learn More

Manganese(III) oxide, 98%

Product Code. 11335039
Change view
Click to view available options
Quantity:
500 g
2 kg
Unit Size:
2kg
500g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11335039 2 kg 2kg
11325039 500 g 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11335039 Supplier Thermo Scientific Alfa Aesar Supplier No. 087791.A3
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 1317-34-6 | Mn2O3 | 157.873 g/mol

Manganese(III) oxide is used to produce ceramic magnets and semiconductors. Further, it serves as a precursor to lithium manganese oxide. In addition to this, it acts as a useful battery cathode.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1317-34-6
Molecular Formula Mn2O3
Molecular Weight (g/mol) 157.873
MDL Number MFCD00016217
InChI Key GEYXPJBPASPPLI-UHFFFAOYSA-N
Synonym manganese iii oxide, manganic oxide, dimanganese trioxide, manganese sesquioxide, manganese trioxide, manganese manganate, manganese sisquioxide, manganese 3+ oxide, manganese oxide, oxo oxomanganiooxy manganese
PubChem CID 14824
IUPAC Name oxo(oxomanganiooxy)manganese
SMILES O=[Mn]O[Mn]=O

Specifications

Melting Point 1080°C (decomposition)
Color Black
Physical Form Powder
Quantity 2 kg
Assay Percent Range 98%
Merck Index 14,5737
Solubility Information Soluble in acid and ammonium chloride. Insoluble in water,alcohol and acetone.
Formula Weight 157.86
Percent Purity 98%
Odor Odorless
Density 4.50 g/mL
Chemical Name or Material Manganese (III) oxide
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.