missing translation for 'onlineSavingsMsg'
Learn More

Manganese(III)acetylacetonate, 97%

Product Code. 15760859 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Packaging:
Glass bottle
Unit Size:
100g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
15760859 100 g Glass bottle 100g
10546821 5 g Glass bottle 5g
10672942 25 g Glass bottle 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 15760859 Supplier Thermo Scientific Acros Supplier No. 343981000
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 14284-89-0 | C15H21MnO6 | 352.27 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 14284-89-0
Molecular Formula C15H21MnO6
Molecular Weight (g/mol) 352.27
MDL Number MFCD00000023
InChI Key HYZQBNDRDQEWAN-LNTINUHCSA-K
Synonym mn iii acetylacetonate, tris acetylacetonato manganese iii
PubChem CID 122130696
IUPAC Name manganese(3+);(E)-4-oxopent-2-en-2-olate
SMILES [Mn+3].C\C([O-])=C\C(C)=O.C\C([O-])=C\C(C)=O.C\C([O-])=C\C(C)=O

Specifications

Melting Point 160°C
Color Black
Elemental Analysis 96% min.
Infrared Spectrum Authentic
Assay Percent Range 97%
Packaging Glass bottle
Quantity 100 g
Fieser 02,264
Formula Weight 352.27
Percent Purity 97%
Physical Form Crystalline Powder
Chemical Name or Material Manganese(III)acetylacetonate
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.