missing translation for 'onlineSavingsMsg'
Learn More

Manganese(III) acetate hydrate, 96%

Product Code. 11377288
Change view
Click to view available options
Quantity:
10 g
50 g
Unit Size:
10g
50g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11377288 10 g 10g
11387288 50 g 50g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11377288 Supplier Thermo Scientific Alfa Aesar Supplier No. 030519.09
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 19513-05-4 | C6H9MnO6 | 232.07 g/mol

It acts as a mild and selective oxidizing agent in organic synthesis and materials science. Manganese(III) Acetate Dihydrate is also a reagent used for radical cyclizations and α-keto-acetoxylation.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 19513-05-4
Molecular Formula C6H9MnO6
Molecular Weight (g/mol) 232.07
MDL Number MFCD00150022
InChI Key AHSBSUVHXDIAEY-UHFFFAOYSA-K
IUPAC Name manganese(3+) triacetate
SMILES [Mn+3].CC([O-])=O.CC([O-])=O.CC([O-])=O

Specifications

Color Pink
Physical Form Crystalline
Quantity 10 g
Linear Formula Mn(OOCCH3)3·xH2O
Solubility Information It is soluble in water.
Formula Weight 232.07 (Anhydrous)
Percent Purity 96%
Sensitivity Hygroscopic
Density 1.59 g/mL
Chemical Name or Material Manganese(III) acetate hydrate
compliance-icons
Product Identifier
  • Manganese(III) acetate hydrate
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P362-Take off contaminated clothing.
Supplemental information
  • MIXTURE LIST-Contain: Manganese(III) acetate hydrate

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.