missing translation for 'onlineSavingsMsg'
Learn More

Manganese(III) 2,4-pentanedionate

Product Code. 11321869
Change view
Click to view available options
Quantity:
10 g
50 g
250 g
Unit Size:
10g
250g
50g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11321869 10 g 10g
11331869 50 g 50g
11341869 250 g 250g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11321869 Supplier Thermo Scientific Alfa Aesar Supplier No. 012530.09
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 14284-89-0 | C15H21MnO6 | 352.27 g/mol

It is mainly used as a solvent in paint and a precursor to vitamin E.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 14284-89-0
Molecular Formula C15H21MnO6
Molecular Weight (g/mol) 352.27
MDL Number MFCD00000023
InChI Key HYZQBNDRDQEWAN-LNTINUHCSA-K
Synonym mn iii acetylacetonate, tris acetylacetonato manganese iii
PubChem CID 122130696
IUPAC Name manganese(3+);(E)-4-oxopent-2-en-2-olate
SMILES [Mn+3].C\C([O-])=C\C(C)=O.C\C([O-])=C\C(C)=O.C\C([O-])=C\C(C)=O

Specifications

Melting Point ∼160°C (decomposition)
Quantity 10 g
Sensitivity Hygroscopic
Solubility Information Soluble in benzene, ethyl acetate
Formula Weight 352.24
Physical Form Powder
Chemical Name or Material Manganese(III) 2,4-pentanedionate

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.