missing translation for 'onlineSavingsMsg'
Learn More

Manganese(II) titanium oxide, 99.9% (metals basis)

Product Code. 11372228
Change view
Click to view available options
Quantity:
10 g
50 g
Unit Size:
10g
50g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11372228 50 g 50g
11382228 10 g 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11372228 Supplier Thermo Scientific Alfa Aesar Supplier No. 013133.18
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 12032-74-5 | MnTiO3

Manganese(II) titanium oxide is used as pharamaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 12032-74-5
Molecular Formula MnTiO3
MDL Number MFCD00016216
Synonym manganese titanium trioxide, manganese ii titanate, manganese 2+ ; oxygen 2-; titanium 4+, titanium 4+ ion manganese 2+ ion trioxidandiide

Specifications

Melting Point 1360°C
Color Yellow
Physical Form Powder
Quantity 50 g
Assay Percent Range 99.9% (Metals basis)
Solubility Information Insoluble in water.
Formula Weight 150.84
Odor Odorless
Chemical Name or Material Manganese(II) titanium oxide

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.