missing translation for 'onlineSavingsMsg'
Learn More

Manganese(II) sulfide, 99.9% (metals basis)

Product Code. 11347567
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Unit Size:
100g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11347567 25 g 25g
11357567 5 g 5g
11337567 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11347567 Supplier Thermo Scientific Alfa Aesar Supplier No. 012835.14
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 18820-29-6 | MnS | 87.00 g/mol

it is used as an intermediate in chemical research& pharmaceutical research.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 18820-29-6
Molecular Formula MnS
Molecular Weight (g/mol) 87.00
MDL Number MFCD00014210
InChI Key VCTOKJRTAUILIH-UHFFFAOYSA-N
Synonym manganese sulfide, manganese ii sulfide, manganous sulfide, manganese sulphide, manganese 2+ sulfide, 12687-82-0 cpd with unspecified mf, manganese ii sulfide, ∼100 mesh particle size
PubChem CID 87809
SMILES [S--].[Mn++]

Specifications

Melting Point 1610°C (decomposition)
Color Green
Physical Form Powder
Quantity 25 g
Assay Percent Range 99.9% (Metal basis)
Solubility Information It is soluble in water. 0.47mg/100 mL (18°C).
IUPAC Name manganese(2+) sulfanediide
Formula Weight 87
Appearance Green powder
Density 3.99
Chemical Name or Material Manganese(II) sulfide
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.