missing translation for 'onlineSavingsMsg'
Learn More

Manganese(II) sulfate tetrahydrate, 99% (metals basis)

Product Code. 11499577
Change view
Click to view available options
Quantity:
100 g
500 g
Unit Size:
100g
500g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11499577 500 g 500g
11489577 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11499577 Supplier Thermo Scientific Alfa Aesar Supplier No. B22081.36
  • ONLINE EXCLUSIVE PRICE
    £127.00
    £114.65 / 500g
    Save £12.35
    9% Off
 Restricted Item
In Stock
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 10101-68-5 | H8MnO8S | 223.054 g/mol

Manganese(II) sulfate tetrahydrate is an essential mineral used in capsule, tablet and liquid form, which provides essential nutrients like vitamin, protein and in similar nutritional substance. It undergoes electrolysis to give manganese dioxide. Upon oxidation, it gives chemical manganese dioxide (CMD) and finds application in dry- cell batteries.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Manganese(II) sulfate tetrahydrate is an essential mineral used in capsule, tablet and liquid form, which provides essential nutrients like vitamin, protein and in similar nutritional substance. It undergoes electrolysis to give manganese dioxide. Upon oxidation, it gives chemical manganese dioxide (CMD) and finds application in dry- cell batteries.

Solubility
Soluble in water.

Notes
Loses all water at 400-450°. Hygroscopic. Incompatible with strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 10101-68-5
Molecular Formula H8MnO8S
Molecular Weight (g/mol) 223.054
MDL Number MFCD00149160
InChI Key CDUFCUKTJFSWPL-UHFFFAOYSA-L
Synonym manganese sulfate tetrahydrate, manganese ii sulfate tetrahydrate, unii-f46lh60l4m, manganese sulphate, tetrahydrate, mnso4.4h2o, acmc-1c6k2, mn.so4.4h2o, manganese 2+ sulfate tetrahydrate, manganese ii sulphate tetrahydrate
PubChem CID 22572548
ChEBI CHEBI:86358
IUPAC Name manganese(2+);sulfate;tetrahydrate
SMILES O.O.O.O.[O-]S(=O)(=O)[O-].[Mn+2]

Specifications

Quantity 500 g
Linear Formula MnSO4·4H2O
UN Number UN3077
Solubility Information Soluble in water.
Formula Weight 223.06 (151.00 Anhydrous)
Percent Purity 99%
Sensitivity Hygroscopic
Assay (metals basis)
Chemical Name or Material Manganese(II) sulfate tetrahydrate
compliance-icons compliance-icons compliance-icons
Product Identifier
  • Manganese(II) sulfate tetrahydrate
Signal Word
  • Danger
Hazard Category
  • LONG-TERM AQUATIC HAZARD Chronic 2
  • Serious eye damage/eye irritation Category 1
  • Specific target organ toxicity after repeated exposure Category 2
Hazard Statement
  • H318-Causes serious eye damage.
  • H373-May cause damage to organs.
  • H411-Toxic to aquatic life with long lasting effects.
Precautionary Statement
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P273-Avoid release to the environment.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
Supplemental information
  • MIXTURE LIST-Contain: Manganese(II) sulfate tetrahydrate, Manganese(II) sulfate

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.