missing translation for 'onlineSavingsMsg'
Learn More

Manganese(II) sulfate monohydrate, ACS, 98.0-101.0%

Product Code. 11367668
Change view
Click to view available options
Quantity:
500 g
2 kg
Unit Size:
2kg
500g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11367668 2 kg 2kg
11357668 500 g 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11367668 Supplier Thermo Scientific Alfa Aesar Supplier No. 033341.A3
  • ONLINE EXCLUSIVE PRICE
    £113.22
    £92.65 / 2kg
    Save £20.57
    18% Off
 Restricted Item
Estimated Shipment: 03-12-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 10034-96-5 | H2MnO5S | 169.01 g/mol

Manganese(II) sulfate monohydrate is used as a colorant in dyes, fertilizers, animal feeds and red glazes on porcelain. Further, it is used in paints, ceramics, nutrient and dietary supplement. It is involved in the preparation of manganese dioxide. In addition, it serves as a precursor to manganese metal and other manganese compounds.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 10034-96-5
Molecular Formula H2MnO5S
Molecular Weight (g/mol) 169.01
MDL Number MFCD00149159
InChI Key ISPYRSDWRDQNSW-UHFFFAOYSA-L
Synonym manganese sulfate monohydrate, manganese ii sulfate monohydrate, manganese sulfate hydrate, manganese ii sulfate hydrate, mnso4.h2o, manganous sulfate monohydrate, unii-w00lys4t26, manganese 2+ sulfate monohydrate, manganese sulfate usp, manganese 2+ sulfate hydrate
PubChem CID 177577
ChEBI CHEBI:86364
IUPAC Name manganese(2+);sulfate;hydrate
SMILES O.[Mn++].[O-]S([O-])(=O)=O

Specifications

Color Pink
Physical Form Powder
Quantity 2 kg
Appearance Pink powder
Assay Percent Range 98.0-101.0%
Density 2.95
Solubility Information Soluble in water and alcohol. Insoluble in ether.
Formula Weight 169.02 (151.00anhydrous)
Grade ACS
Chemical Name or Material Manganese(II) sulfate monohydrate

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.