missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Manganese(II) sulfate monohydrate, ACS, 98.0-101.0%, Thermo Scientific Chemicals
Click to view available options
Quantity:
500 g
2 kg
Description
Manganese(II) sulfate monohydrate is used as a colorant in dyes, fertilizers, animal feeds and red glazes on porcelain. Further, it is used in paints, ceramics, nutrient and dietary supplement. It is involved in the preparation of manganese dioxide. In addition, it serves as a precursor to manganese metal and other manganese compounds.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Color | Pink |
Physical Form | Powder |
Quantity | 500 g |
Assay Percent Range | 98.0-101.0% |
Solubility Information | Soluble in water and alcohol. Insoluble in ether. |
Formula Weight | 169.02 (151.00 Anhydrous) |
Grade | ACS |
Appearance | Pink powder |
Density | 2.95 |
Chemical Name or Material | Manganese(II) sulfate monohydrate |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'