missing translation for 'onlineSavingsMsg'
Learn More

Manganese(II) sulfate monohydrate, 99%

Product Code. 11488237
Change view
Click to view available options
Quantity:
100 g
500 g
2500 g
Unit Size:
100g
2500g
500g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11488237 500 g 500g
11478237 100 g 100g
11498237 2500 g 2500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11488237 Supplier Thermo Scientific Alfa Aesar Supplier No. A17615.36
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 10034-96-5 | H2MnO5S | 169.01 g/mol

Manganese(II) sulfate monohydrate acts as a colorant in dyes. It is used in the preparation of manganese and manganese dioxide. It finds application for remediation of manganese-deficient soil. It is also used in dry-cell batteries.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Manganese(II) sulfate monohydrate acts as a colorant in dyes. It is used in the preparation of manganese and manganese dioxide. It finds application for remediation of manganese-deficient soil. It is also used in dry-cell batteries.

Solubility
Soluble in water and alcohol. Insoluble in ether.

Notes
Loses all water at 400-450°. Hygroscopic. Incompatible with strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 10034-96-5
Molecular Formula H2MnO5S
Molecular Weight (g/mol) 169.01
MDL Number MFCD00149159
InChI Key ISPYRSDWRDQNSW-UHFFFAOYSA-L
Synonym manganese sulfate monohydrate, manganese ii sulfate monohydrate, manganese sulfate hydrate, manganese ii sulfate hydrate, mnso4.h2o, manganous sulfate monohydrate, unii-w00lys4t26, manganese 2+ sulfate monohydrate, manganese sulfate usp, manganese 2+ sulfate hydrate
PubChem CID 177577
ChEBI CHEBI:86364
IUPAC Name manganese(2+);sulfate;hydrate
SMILES O.[Mn++].[O-]S([O-])(=O)=O

Specifications

pH 3 to 3.5
Quantity 500 g
UN Number UN3077
Merck Index 14,5739
Solubility Information Soluble in water and alcohol. Insoluble in ether.
Formula Weight 169.02 (151.00 Anhydrous)
Percent Purity 99%
Odor Odorless
Sensitivity Hygroscopic
Density 2.95
Chemical Name or Material Manganese(II) sulfate monohydrate
Show More Show Less
compliance-icons compliance-icons compliance-icons
Product Identifier
  • Manganese(II) sulfate monohydrate
Signal Word
  • Danger
Hazard Category
  • LONG-TERM AQUATIC HAZARD Chronic 2
  • Serious eye damage/eye irritation Category 1
  • Specific target organ toxicity after repeated exposure Category 2
Hazard Statement
  • H318-Causes serious eye damage.
  • H373-May cause damage to organs.
  • H411-Toxic to aquatic life with long lasting effects.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
Supplemental information
  • MIXTURE LIST-Contain: Manganese sulfate monohydrate

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.