missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Manganese(II) sulfate monohydrate, 97%, Thermo Scientific Chemicals
75.33 GBP valid until 2024-03-29
Use promo code "21615" to get your promotional price.
Use promo code "21615" to get your promotional price.
Click to view available options
Quantity:
500 g
2500 g
Description
Manganese(II) sulfate is used as a colorant in dyes. It is also used as feed additives, catalyst for the synthesis of chlorophyll, make plants grow well and increase production. It is used for making dryer of ink and paint. It is used in paper making , porcelain/ceramic, medicine and other industries.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
Manganese(II) sulfate is used as a colorant in dyes. It is also used as feed additives, catalyst for the synthesis of chlorophyll, make plants grow well and increase production. It is used for making dryer of ink and paint. It is used in paper making , porcelain/ceramic, medicine and other industries.
Solubility
Soluble in water. 5-10 g/100 mL at 21°C
Notes
Loses all water at 400-450°
Manganese(II) sulfate is used as a colorant in dyes. It is also used as feed additives, catalyst for the synthesis of chlorophyll, make plants grow well and increase production. It is used for making dryer of ink and paint. It is used in paper making , porcelain/ceramic, medicine and other industries.
Solubility
Soluble in water. 5-10 g/100 mL at 21°C
Notes
Loses all water at 400-450°
Specifications
pH | 3 to 3.5 |
Quantity | 2500 g |
UN Number | UN3077 |
Merck Index | 14,5739 |
Solubility Information | Soluble in water. 5-10g/100 mL at 21°C |
IUPAC Name | manganese(2+) hydrate sulfate |
Formula Weight | 169.02 (151.00 Anhydrous) |
Percent Purity | 97% |
Odor | Odorless |
Sensitivity | Hygroscopic |
Show More |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'