missing translation for 'onlineSavingsMsg'
Learn More

Manganese(II) Sulfate Hydrate, TRC

Product Code. 30472435
Change view
Click to view available options
Quantity:
1 g
5 g
10 g
Unit Size:
10g
1g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30472435 1 g 1g
30465932 10 g 10g
30480529 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30472435 Supplier Toronto Research Chemicals Supplier No. TRCM2458151G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 15244-36-7
Molecular Formula SO42 . x H2O . Mn
Molecular Weight (g/mol) 169.02
Synonym C.I. 77752, Manganese Sulfate (MnSO4) Tetrahydrate, Manganese Sulfate Tetrahydrate, Manganese(2+) Sulfate Tetrahydrate, Manganous Sulfate Tetrahydrate
IUPAC Name manganese;sulfuric acid;hydrate
SMILES O.[Mn].OS(=O)(=O)O

Specifications

Quantity 1 g
InChI Formula InChI=1S/Mn.H2O4S.H2O/c;1-5(2,3)4;/h;(H2,1,2,3,4);1H2
Formula Weight 170.916
Chemical Name or Material Manganese(II) Sulfate Hydrate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.