missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Manganese(II) perchlorate hexahydrate, Reagent Grade, Thermo Scientific Chemicals
29.40 GBP valid until 2024-03-29
Use promo code "21615" to get your promotional price.
Use promo code "21615" to get your promotional price.
Click to view available options
Quantity:
100 g
500 g
Description
It used in rocket fuel. This is one of the most proven materials for etching of liquid crystal displays and critical electronics applications as well as ore extraction. It is also used for electropolishing/etching of aluminum, molybdenum, and other metals.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Melting Point | 165°C (decomposition) |
Physical Form | Crystalline |
Quantity | 100 g |
UN Number | UN1481 |
Solubility Information | Soluble in benzene, ethyl acetate |
Formula Weight | 361.93 (253.84 Anhydrous) |
Grade | Reagent |
Odor | Odorless |
Sensitivity | Hygroscopic |
Chemical Name or Material | Manganese(II) perchlorate hexahydrate |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'