missing translation for 'onlineSavingsMsg'
Learn More

Manganese(II) perchlorate hexahydrate, Reagent Grade

Product Code. 11351669
Change view
Click to view available options
Quantity:
100 g
500 g
Unit Size:
100g
500g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11351669 500 g 500g
11361669 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11351669 Supplier Thermo Scientific Alfa Aesar Supplier No. 012309.36
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 15364-94-0 | Cl2H12MnO14 | 361.92 g/mol

It used in rocket fuel. This is one of the most proven materials for etching of liquid crystal displays and critical electronics applications as well as ore extraction. It is also used for electropolishing/etching of aluminum, molybdenum, and other metals.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 15364-94-0
Molecular Formula Cl2H12MnO14
Molecular Weight (g/mol) 361.92
MDL Number MFCD00150255
InChI Key AIFGVOMWPFMOCN-UHFFFAOYSA-L
Synonym manganese perchlorate hexahydrate, manganese ii perchlorate hexahydrate, perchloric acid, manganese 2+ salt, hexahydrate, 2clo4.mn.6h2o, manganese 2+ diperchlorate hexahydrate, manganese 2+ ion hexahydrate diperchlorate, manganese ii hexahydrate diperchlorate ion, manganese ii perchlorate hexahydrate, reagent grade
PubChem CID 203855
IUPAC Name manganese(2+);diperchlorate;hexahydrate
SMILES O.O.O.O.O.O.[Mn++].[O-][Cl](=O)(=O)=O.[O-][Cl](=O)(=O)=O

Specifications

Melting Point 165°C (decomposition)
Physical Form Crystalline
Quantity 500 g
UN Number UN1481
Solubility Information Soluble in benzene, ethyl acetate
Formula Weight 361.93 (253.84 Anhydrous)
Grade Reagent
Odor Odorless
Sensitivity Hygroscopic
Chemical Name or Material Manganese(II) perchlorate hexahydrate

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.