missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Manganese(II) perchlorate hexahydrate, 99.995% (metals basis), Thermo Scientific Chemicals
Click to view available options
Quantity:
10 g
50 g
250 g
Description
Manganese(II) perchlorate hexahydrate, is a good oxidizing agent and also acts as an intermediates in chemical research.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Melting Point | 165°C (decomposition) |
Physical Form | Crystalline |
Quantity | 10 g |
UN Number | UN1481 |
Solubility Information | Soluble in water,benzene,ethyl acetate |
Formula Weight | 361.93 (253.84 Anhydrous) |
Percent Purity | 99.995% |
Odor | Odorless |
Sensitivity | Hygroscopic |
Assay | (metals basis) |
Show More |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'