Learn More
Manganese(II) oxide, Mn 76.0-78.0%, typically 99%, Thermo Scientific Chemicals
Use promo code "21615" to get your promotional price.
Description
The main use of manganese dioxide is in dry-cell batteries, such as the alkaline battery and the zinc-carbon battery. It is useful as a pigment, and as a precursor to other manganese compounds. It is used as a reagent in organic synthesis, for instance, for the oxidation of allylic alcohols and arylmethyl alcohols. It is also employed in various industrial purposes such as in adhesives and sealant chemicals, lubricants and lubricant additives, fertilizers, oxidizing/reducing agents, plating agents and surface treating agents, processing aids, air care products, paints, and coatings.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Color | Green |
Physical Form | Powder |
Quantity | 500 g |
Assay Percent Range | Mn 76.0-78.0%, typically 99% |
Solubility Information | Insoluble in water. Soluble in hydrochloric acid. |
Formula Weight | 70.94 |
Odor | Odorless |
Appearance | Green powder |
Density | 5 g/cm3 at 20°C |
Chemical Name or Material | Manganese(II) oxide, Mn 76.0-78.0% |
RUO – Research Use Only