missing translation for 'onlineSavingsMsg'
Learn More

Manganese(II) oxide, 99.99% (metals basis)

Product Code. 11470740
Change view
Click to view available options
Quantity:
10 g
50 g
Unit Size:
10g
50g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11470740 50 g 50g
11460740 10 g 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11470740 Supplier Thermo Scientific Alfa Aesar Supplier No. 044441.18
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 1344-43-0 | MnO | 70.94 g/mol

The main use of manganese dioxide is in dry-cell batteries, such as the alkaline battery and the zinc-carbon battery. It is useful as a pigment, and as a precursor to other manganese compounds. It is used as a reagent in organic synthesis, for instance, for the oxidation of allylic alcohols and arylmethyl alcohols. It is also employed in various industrial purposes such as adhesives and sealant chemicals, lubricants and lubricant additives, fertilizers, oxidizing/reducing agents, plating agents and surface treating agents, processing aids, air care products, paints and coatings.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1344-43-0
Molecular Formula MnO
Molecular Weight (g/mol) 70.94
MDL Number MFCD00016226
InChI Key VASIZKWUTCETSD-UHFFFAOYSA-N
Synonym manganese ii oxide, manganosite, manganese monoxide, manganese green, cassel green, manganese oxide, rosensthiel green, nu-manese, manganese monooxide, manganese protoxide
PubChem CID 14940
IUPAC Name oxomanganese
SMILES O=[Mn]

Specifications

Color Green
Physical Form Chunks and/or Powder
Quantity 50 g
Solubility Information Soluble in hydrochloric acid. Insoluble in water.
Formula Weight 70.94
Percent Purity 99.99%
Odor Odorless
Density 5.0 g/mL
Assay (metals basis)
Chemical Name or Material Manganese (II) oxide

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.