missing translation for 'onlineSavingsMsg'
Learn More

Manganese(II) oxalate dihydrate, Mn 30% min

Product Code. 11336998
missing translation for 'changeView'
missing translation for 'orderingAttributeHoverText'
Quantity:
5 g
50 g
250 g
1 kg
missing translation for 'unitSize'
1kg
250g
50g
5g
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11336998 50 g 50g
11306998 5 g 5g
11326998 250 g 250g
11316998 1 kg 1kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 missing translation for 'options'
This item is not returnable. View return policy
Product Code. 11336998 missing translation for 'mfr' Thermo Scientific Alfa Aesar missing translation for 'supplierNo' 014685.18
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

As a paint and varnish drier | CAS: 6556-16-7 | C2H4MnO6 | 178.986 g/mol

Manganese(II) oxalate dihydrate is used as a paint and varnish drier. It is also used as a chemical reagent as well as to prepare other manganese compounds. In addition, it is used as a semiconductor photosensitive material.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 6556-16-7
Molecular Formula C2H4MnO6
Molecular Weight (g/mol) 178.986
MDL Number MFCD00150452
InChI Key HDJUVFZHZGPHCQ-UHFFFAOYSA-L
Synonym manganese ii oxalate 2-hydrate
PubChem CID 131882857
IUPAC Name manganese(2+);oxalate;dihydrate
SMILES C(=O)(C(=O)[O-])[O-].O.O.[Mn+2]

Specifications

Melting Point 100°C (decomposition)
Color White
Density 2.453 g/mL
Odor Odorless
Quantity 50 g
Merck Index 14,5734
Solubility Information Soluble in dilute acids; Slightly soluble in water.
Formula Weight 178.99 (142.96 Anhydrous)
Concentration or Composition (by Analyte or Components) Mn 30% min.
Physical Form Chunks and/or Powder
Chemical Name or Material Manganese(II) oxalate dihydrate
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.