missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Manganese(II) nitrate, 50% w/w aq. soln., Thermo Scientific Chemicals
Click to view available options
Quantity:
50 g
500 g
2 kg
Description
Manganese(II) nitrate is used as a precursor to prepare the oxides of manganese. It is involved in the preparation of electronic components. Further, it is utilized for the phosphorous treatment of metal surface.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Color | Pink |
Physical Form | Liquid |
Quantity | 500 g |
Solubility Information | Miscible with water. |
Formula Weight | 178.95 |
Appearance | Pink liquid |
Vapor Pressure | 23 hPa (17mm Hg) at 20°C |
Chemical Name or Material | Manganese(II) nitrate, 50% w/w aq. Soln |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'