missing translation for 'onlineSavingsMsg'
Learn More

Manganese(II) nitrate, 50% w/w aq. soln.

Product Code. p-4407634
Click to view available options
Quantity:
50 g
500 g
2 kg
Unit Size:
2kg
500g
50g
This item is not returnable. View return policy

Product Code. 11337668

Brand: Thermo Scientific Alfa Aesar 033340.18

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

CAS: 10377-66-9 | MnN2O6 | 178.946 g/mol

Manganese(II) nitrate is used as a precursor to prepare the oxides of manganese. It is involved in the preparation of electronic components. Further, it is utilized for the phosphorous treatment of metal surface.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 10377-66-9
Molecular Formula MnN2O6
Molecular Weight (g/mol) 178.946
MDL Number MFCD00011112
InChI Key MIVBAHRSNUNMPP-UHFFFAOYSA-N
Synonym manganese nitrate, manganous nitrate, manganese dinitrate, manganous dinitrate, manganese ii nitrate, manganese 2+ nitrate, nitric acid, manganese 2+ salt, manganese ii nitrate, anhydrous, manganesenitrate
PubChem CID 61511
IUPAC Name manganese(2+);dinitrate
SMILES [N+](=O)([O-])[O-].[N+](=O)([O-])[O-].[Mn+2]

Specifications

Color Pink
Physical Form Liquid
Quantity 50 g
Formula Weight 178.95
Appearance Pink liquid
Vapor Pressure 23 hPa (17mm Hg) at 20°C
Chemical Name or Material Manganese(II) nitrate, 50% w/w aq. Soln

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.