missing translation for 'onlineSavingsMsg'
Learn More

Manganese(II) methoxide

Product Code. 11338067
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11338067 25 g 25g
11318067 1 g 1g
11328067 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11338067 Supplier Thermo Scientific Alfa Aesar Supplier No. 022563.14
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 7245-20-7 | C2H6MnO2 | 117.006 g/mol

Manganese(II) methoxide is used as a biochemical for proteomics research and it is also used in convenient for carboxylating active methylene compounds.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7245-20-7
Molecular Formula C2H6MnO2
Molecular Weight (g/mol) 117.006
MDL Number MFCD00061476
InChI Key UQIRCVPINUNHQY-UHFFFAOYSA-N
Synonym manganese ii methoxide, acmc-20akkm, dimethoxymanganese ii, manganese ii bis methoxide, manganese 2+ bis methoxide, manganese 2+ ion bis methoxide
PubChem CID 10219413
IUPAC Name manganese(2+);methanolate
SMILES C[O-].C[O-].[Mn+2]

Specifications

Linear Formula Mn(OCH3)2
Quantity 25 g
UN Number UN3181
Sensitivity Moisture Sensitive
Solubility Information Soluble in methanol
Formula Weight 117.01
Physical Form Powder
Chemical Name or Material Manganese(II) methoxide

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.