missing translation for 'onlineSavingsMsg'
Learn More

Manganese(II) iodide, ultra dry, 99.99% (metals basis)

Product Code. 11318557
Change view
Click to view available options
Quantity:
2 g
10 g
Packaging:
Ampouled under argon
Plastic bottle; Ampouled under argon
Unit Size:
10g
2g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
11318557 2 g Ampouled under argon 2g
11308557 10 g Plastic bottle; Ampouled under argon 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11318557 Supplier Thermo Scientific Alfa Aesar Supplier No. 035710.04
 Restricted Item
Estimated Shipment: 27-11-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Often used in the lighting industry and it is a source of its derivatives. | CAS: 7790-33-2 | I2Mn

It is often used in the lighting industry and it is a source of its derivatives.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7790-33-2
Molecular Formula I2Mn
MDL Number MFCD00016225

Specifications

Melting Point 638°C
Physical Form Powder or Flakes
Quantity 2 g
Packaging Ampouled under argon
Sensitivity Air, light, and moisture sensitive
Specific Gravity 5
Density 5
Linear Formula MnI2
Solubility Information Soluble in water.
Formula Weight 308.75
Percent Purity 99.99%
Chemical Name or Material Manganese(II) iodide
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.