missing translation for 'onlineSavingsMsg'
Learn More

Manganese(II) fluoride, 99%

Product Code. 11397207
Click to view available options
Quantity:
10 g
50 g
Unit Size:
10g
50g
This item is not returnable. View return policy

Product Code. 11397207

Brand: Thermo Scientific Alfa Aesar 011527.09

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 7782-64-1 | F2Mn | 92.935 g/mol

Manganese(II) fluoride is used in the manufacture of special kinds of glass and lasers.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7782-64-1
Molecular Formula F2Mn
Molecular Weight (g/mol) 92.935
MDL Number MFCD00016222
InChI Key CTNMMTCXUUFYAP-UHFFFAOYSA-L
Synonym manganese ii fluoride, manganese fluoride mnf2, manganese fluorure french, manganese fluorure, mnf2, manganese fluoride di, manganese ii fluoride, anhydrous trace metals basis
PubChem CID 24528
IUPAC Name difluoromanganese
SMILES F[Mn]F

Specifications

Melting Point 856°C
Color Red
Physical Form Crystalline Powder
Quantity 10 g
Assay Percent Range 99%
Solubility Information Insoluble in water,alcohol. Soluble in dilute HF,conc. HCl or HNO3
Formula Weight 92.93
Sensitivity Hygroscopic
Density 3.98
Chemical Name or Material Manganese(II) fluoride

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.