missing translation for 'onlineSavingsMsg'
Learn More

Manganese(II) chloride, ultra dry, 99.998% (metals basis)

Product Code. 11307008
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11307008 5 g 5g
11396998 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11307008 Supplier Thermo Scientific Alfa Aesar Supplier No. 014697.06
 Restricted Item
Estimated Shipment: 07-08-2026
Add to basket
This item is not returnable. View return policy

CAS: 7773-01-5 | Cl2Mn | 125.84 g/mol

Manganese(II) chloride is used in formation of an active Mn(0) species useful for radical cyclization reactions. It is used as catalyst, raw material for paint drier and antiknock agent for gasoline. It is also used in the production of pigments, dyes, car batteries, chemical reagents and trace elements.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

  • Ampouled under argon
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7773-01-5
Molecular Formula Cl2Mn
Molecular Weight (g/mol) 125.84
MDL Number MFCD00011114
InChI Key GLFNIEUTAYBVOC-UHFFFAOYSA-L
Synonym manganese chloride, manganese ii chloride, manganese 2+ ion dichloride, manganesechloride, acmc-20akkt, manganous chloride,anhydrous, ksc171o6h, manganous chloride, anhydr, manganese ii chloride anhydrous crystalline
PubChem CID 10313134
ChEBI CHEBI:63041
IUPAC Name manganese(2+);dichloride
SMILES [Cl-].[Cl-].[Mn++]

Specifications

Boiling Point 1190°C
Melting Point 650°C
Color Pink
Physical Form Beads
Quantity 5 g
Linear Formula MnCl2
UN Number UN3288
Merck Index 14,5728
Solubility Information Soluble in water(723g/L at 25 C)
Formula Weight 125.84
Percent Purity 99.998%
Odor Odorless
Packaging Ampule
Sensitivity Hygroscopic
Density 2.977 g/mL
Assay (metals basis)
Chemical Name or Material Manganese(II) chloride, Ultra dry
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.