missing translation for 'onlineSavingsMsg'
Learn More

Manganese(II) chloride, ultra dry, 99.99% (metals basis)

Product Code. 11310068
Click to view available options
Quantity:
2 g
10 g
50 g
Unit Size:
10g
2g
50g
This item is not returnable. View return policy

Product Code. 11310068

Brand: Thermo Scientific Alfa Aesar 042844.18

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

Denne vare kan ikke returneres. Se returpolicy

Plays an important role in the production of dry cell batteries. | CAS: 7773-01-5 | Cl2Mn | 125.84 g/mol

Manganese(II) chloride, plays an important role in the production of dry cell batteries. It is also used as a starting material to the methylcyclopentadienyl manganese tricarbonyl, which has antiknock property. Further, it is used in the formation of an active Manganese(0) species useful for radical cyclization. It is also used in 31P-NMR to determine the size and lamellarity of phospholipidvesicles reactions.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Kemiske identifikatorer

CAS 7773-01-5
Molecular Formula Cl2Mn
Molecular Weight (g/mol) 125.84
MDL Number MFCD00011114
InChI Key GLFNIEUTAYBVOC-UHFFFAOYSA-L
Synonym manganese chloride, manganese ii chloride, manganese 2+ ion dichloride, manganesechloride, acmc-20akkt, manganous chloride,anhydrous, ksc171o6h, manganous chloride, anhydr, manganese ii chloride anhydrous crystalline
PubChem CID 10313134
ChEBI CHEBI:63041
IUPAC Name manganese(2+);dichloride
SMILES [Cl-].[Cl-].[Mn++]

Tekniske data

Boiling Point 1190°C
Melting Point 650°C
Physical Form Powder
Quantity 50 g
Linear Formula MnCl2
UN Number UN3288
Solubility Information Soluble in water,pyridine and alcohol. Insoluble in ether.
Formula Weight 125.84
Percent Purity 99.99%
Packaging Ampouled under argon
Sensitivity Hygroscopic
Specific Gravity 2.977
Density 2.977
Chemical Name or Material Manganese(II) chloride
Vis mere Vis mindre

RUO – Research Use Only

Korrektion af produktindhold

Dit input er vigtigt for os. Udfyld denne formular for at give feedback relateret til indholdet på dette produkt.

Produkttitel

Ved at klikke på Send, anerkender du, at du kan blive kontaktet af Fisher Scientific med hensyn til den feedback, du har givet i denne formular. Vi deler ikke dine oplysninger til andre formål. Alle angivne kontaktoplysninger skal også vedligeholdes i overensstemmelse med vores Privatlivspolitik.

Mange tak! Din feedback er blevet sendt. Hos Fisher Scientific arbejder vi altid på at forbedre vores indhold for dig. Vi sætter pris på din feedback.