missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Manganese(II) chloride, ultra dry, 99.99% (metals basis), Thermo Scientific Chemicals
Click to view available options
Quantity:
5 g
25 g
100 g
Description
Manganese(II) chloride is used in formation of an active Mn(0) species useful for radical cyclization reactions. It is used as catalyst, raw material for paint drier and antiknock agent for gasoline. It iis also used in the production of pigments, dyes, car batteries, chemical reagents and trace elements.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
- Ampouled under argon
Specifications
Boiling Point | 1190°C |
Melting Point | 650°C |
Color | Pink |
Physical Form | Beads |
Quantity | 25 g |
Linear Formula | MnCl2 |
UN Number | UN3288 |
Merck Index | 14,5728 |
Solubility Information | Soluble in water(723g/L at 25 C) |
IUPAC Name | manganese(2+);dichloride |
Show More |
Safety and Handling
Signal Word
- Danger
- Acute toxicity Category 3
- Serious eye damage/eye irritation Category 1
- Specific target organ toxicity after repeated exposure Category 2
- H301-Toxic if swallowed.
- H318-Causes serious eye damage.
- H373-May cause damage to organs.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P310-Immediately call a POISON CENTER/doctor/.
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'