missing translation for 'onlineSavingsMsg'
Learn More

Manganese(II) chloride tetrahydrate, ACS, 98.0-101.0%

Product Code. p-4781992
Click to view available options
Quantity:
100 g
500 g
2 kg
Unit Size:
100g
2kg
500g
This item is not returnable. View return policy

Product Code. 11317978

Brand: Thermo Scientific Alfa Aesar 036526.22

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 13446-34-9 | Cl2H8MnO4 | 197.90 g/mol

It is used in the production of dry cell batteries. It is a precursor material for antiknock compound methylcyclopentadienyl manganese tricarbony. It is also useful for coupling reactions as well as organic synthesis. It is a catalyst for fast and slow dehydrogenation of ball milled lithium alanate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 13446-34-9
Molecular Formula Cl2H8MnO4
Molecular Weight (g/mol) 197.90
MDL Number MFCD00149792
InChI Key CNFDGXZLMLFIJV-UHFFFAOYSA-L
IUPAC Name manganese(2+) tetrahydrate dichloride
SMILES O.O.O.O.[Cl-].[Cl-].[Mn++]

Specifications

Melting Point 58°C
Color Pink
Physical Form Crystalline
Quantity 100 g
Assay Percent Range 98-101%
Solubility Information Soluble in water,pyridine and ethanol. Insoluble in ether.
Formula Weight 197.90 (125.84anhydrous)
Grade ACS
Odor Odorless
Sensitivity Hygroscopic
Appearance Pink crystalline
Density 2.01
Chemical Name or Material Manganese(II) chloride tetrahydrate
Show More Show Less

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.