missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Manganese(II) Chloride Tetrahydrate, ≥99%, Reagent Grade, Honeywell™
Shop All Honeywell Research Chemicals Products
Click to view available options
Quantity:
100 g
500 g
Packaging:
Poly bottle
Unit Size:
100g
500g
Description
- ≥95% for general laboratory use.
Specifications
Specifications
| Melting Point | 58°C |
| Color | Pink |
| pH | 4.0 to 6.0 (at 20°C) |
| Physical Form | Crystalline |
| Quantity | 100 g |
| Linear Formula | MnCl2 · 4H2O |
| UN Number | NONH for all modes of transport |
| Solubility Information | Completely soluble in water |
| Formula Weight | 197.91g/mol |
| Percent Purity | ≥99% |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction