missing translation for 'onlineSavingsMsg'
Learn More

Manganese(II) chloride tetrahydrate, 99% (metals basis)

Product Code. 11357287
Change view
Click to view available options
Quantity:
100 g
500 g
Unit Size:
100g
500g
Product Code. Quantity unitSize
11357287 500 g 500g
11347287 100 g 100g
2 options
This item is not returnable. View return policy

Product Code. 11357287

Brand: Thermo Scientific Alfa Aesar 011563.36

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 13446-34-9 | Cl2H8MnO4 | 197.90 g/mol

Manganese(II) chloride tetrahydrate is used in the tailing of DNA - MnCl2 buffer is recommended for dC and dG tailing and in reverse transcription of RNA with thermostable DNA polymerase.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 13446-34-9
Molecular Formula Cl2H8MnO4
Molecular Weight (g/mol) 197.90
MDL Number MFCD00149792
InChI Key CNFDGXZLMLFIJV-UHFFFAOYSA-L
IUPAC Name manganese(2+) tetrahydrate dichloride
SMILES O.O.O.O.[Cl-].[Cl-].[Mn++]

Specifications

Melting Point 58°C
Color Pink
Physical Form Crystalline
Quantity 500 g
Assay Percent Range 99% (Metal basis)
Solubility Information Soluble in pyridine,ethanol. Insoluble in ether.
Formula Weight 197.90 (125.84 Anhydrous)
Odor Odorless
Sensitivity Hygroscopic
Appearance Pink Crystalline/powder
Density 2.01
Chemical Name or Material Manganese(II) chloride tetrahydrate
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.