missing translation for 'onlineSavingsMsg'
Learn More

Manganese(Ii) Chloride Tetrahydrate, 99.99% trace metals basis, Honeywell™

Product Code. p-7120544 Shop All Honeywell Research Chemicals Products
Click to view available options
Quantity:
5 g
25 g
100 g
Packaging:
Glass Bottle
Poly Bottle
Unit Size:
100g
25g
5g
This item is not returnable. View return policy

Product Code. 15651750

Brand: Honeywell Research Chemicals 2037345G

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

99.99% trace metals basis

  • We are your supplier of high-quality lab reagents and chemicals for analytical and chemistry laboratories:
    Solvents – ACS specific use (HPLC), ACS general use, solvents for histology, and reagent grade for chemical synthesis and other industrial applications.
    Inorganics – Chemical synthesis and inorganic chemistry including essential acids and bases, salts, metals and elements, and reagents for chemical reactions.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 13446-34-9
Molecular Formula Cl2H8MnO4
Molecular Weight (g/mol) 197.90
MDL Number MFCD00149792
InChI Key CNFDGXZLMLFIJV-UHFFFAOYSA-L
IUPAC Name manganese(2+) tetrahydrate dichloride
SMILES O.O.O.O.[Cl-].[Cl-].[Mn++]

Specifications

Melting Point 58°C
Color Pink
Quantity 5 g
Linear Formula MnCl2 · 4H2O
UN Number NONH for all modes of transport
Formula Weight 197.91g/mol
Percent Purity 99.99% (trace metals basis)
Packaging Glass Bottle
Appearance Crystals
Density 1.913g/cu.cm
Chemical Name or Material Manganese(Ii) Chloride Tetrahydrate
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.